Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FcΕRI α subunit Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 06725IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
06-725-IMI 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 06-725-IMI Supplier MilliporeSigma Supplier No. 06725I
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect FcΕRI α subunit using this Anti-FcΕRI α subunit Antibody validated for use in Western Blotting, Immunoprecipitation, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FcεRI α subunit
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum containing 0.05% sodium azide.
Gene Symbols FCER1A; FCE1A
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to the extracellular domain of human FcεRI α subunit.
Purification Method Unpurified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_001992
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.