Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FCAR/CD89, PE, Clone: 488032, R&D Systems™

Catalog No. FAB3939P
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB3939P 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB3939P Supplier R&D Systems Supplier No. FAB3939P
This item is not returnable. View return policy

Applications:

Flow Cytometry

Specifications

Antigen FCAR/CD89
Applications Flow Cytometry
Classification Monoclonal
Clone 488032
Conjugate R-PE
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. P24071
Gene Alias CD89, CD89 antigen, CD89IgA Fc receptor, Fc alpha receptor, Fc fragment of IgA, receptor for, immunoglobulin alpha Fc receptor
Gene Symbols FCAR
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human FCAR/CD89, Gln22-Lys287, Accession # P24071
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2204
Test Specificity Detects human FCAR/CD89 in direct ELISAs. In direct ELISAs, no cross-reactivity with Fc gamma RIA, RIIA, or RIIIB is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.