Learn More
Description
This gene encodes a protein that plays an important role in the immune response. This protein is a high-affinity Fc-gamma receptor. The gene is one of three related gene family members located on chromosome 1. [provided by RefSeq
Specifications
Specifications
| Antigen | FCGR1A |
| Applications | ELISA, In situ PLA |
| Classification | Monoclonal |
| Clone | 1D3 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant FCGR1A. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | FCGR1A |
| Gene Accession No. | NM_000566 |
| Gene Alias | CD64/CD64A/FCRI/FLJ18345/IGFR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.