Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-FCGR2A Clone: 300000000,

Catalog No. ST1571100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1571100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1571100UG Supplier MilliporeSigma™ Supplier No. ST1571100UG

Mouse Monoclonal Antibody

Specifically detects FCGR2A Clone: 300000000 in Human samples, and it is validated for ELISA, Immuno Blotting

This protein is a member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FCGR2A
Applications ELISA, Immunoblot, Immunoblot
Classification Monoclonal
Clone 300000000
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen A recombinant polypeptide corresponding to amino acids 46-151 of human FCGR2A, expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the FCGR2A protein.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.