Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fc fragment of IgG, low affinity IIb, receptor (CD32), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human FCGR2B protein.
Supplier: Abnova Corporation H00002213D01P
Description
Sequence: MGILSFLPVLATESDWADCKSPQPWGHMLLWTAVLFLAPVAGTPAAPPKAVLKLEPQWINVLQEDSVTLTCRGTHSPESDSIQWFHNGNLIPTHTQPSYRFKANNNDSGEYTCQTGQTSLSDPVHLTVLSEWLVLQTPHLEFQEGETIVLRCHSWKDKPLVKVTFFQNGKSKKFSRSDPNFSIPQANHSHSGDYHCTGNIGYTLYSSKPVTITVQAPSSSPMGIIVAVVTGIAVAAIVAAVVALIYCRKKRISALPGYPECREMGETLPEKPANPTNPDEADKVGAENTITYSLLMHPDALEEPDDQNRISpecifications
Fc fragment of IgG, low affinity IIb, receptor (CD32) | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human FCGR2B protein. | |
FCGR2B | |
CD32/CD32B/FCG2/FCGR2/IGFR2 | |
Rabbit | |
Affinity Purified | |
RUO | |
2213 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_004001 | |
FCGR2B | |
FCGR2B (NP_003992.3, 1 a.a. ∼ 310 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction