Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FCRL5/FCRL3 (CD307e/c) Mouse anti-Human, Alexa Fluor 647, Clone: 307307, BD

Catalog No. BDB564343 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564343 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564343 Supplier BD Biosciences Supplier No. 564343
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 307307 monoclonal antibody specifically binds to Fc receptor-like 5 (FCLR5) and crossreacts with Fc receptor-like 3 (FCLR3). These type I transmembrane glycoproteins have sequences that are homologous to classical Fc γreceptors. FCLR5 is also known as CD307e, FCRH5 (Fc receptor homolog 5) and IRTA2 (immunoglobulin superfamily receptor translocation associated 2) whereas FCLR3 is referred to as CD307c, FCRH3 (Fc Receptor Homolog 3) and IRTA3 (immunoglobulin superfamily receptor translocation associated 3). FCRL5 and FCRL3 are differentially expressed by mature B cell subsets and play key roles in controlling signaling through the B cell receptor.

TRUSTED_SUSTAINABILITY

Specifications

Antigen FCRL5/FCRL3 (CD307e/c)
Applications Flow Cytometry
Classification Monoclonal
Clone 307307
Conjugate Alexa Fluor 647
Description FCRL5/FCRL3; FcRH5/FcRH3; IRTA2/IRTA3
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human FCLR5 Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.