Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-FE65, Clone: 3H6,

Catalog No. 05758MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-758-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-758-MI Supplier MilliporeSigma™ Supplier No. 05758

Specifications

Antigen FE65
Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 3H6
Conjugate Unconjugated
Formulation Immunoaffinity Purified immunoglobulin in 0.2M Tris-glycine, pH 7.2, 0.15M NaCl, 0.05% sodium azide with 5mg/ml BSA
Gene Accession No. NM_001164
Gene Symbols APBB1, RIR, FE65, MGC:9072, Fe65
Host Species Mouse
Immunogen GST fusion protein corresponding to amino acids 254-293 of mouse FE65, WW domain
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.