Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGF-2/basic FGF Mouse anti-Human, Bovine, Mouse, Rat, Clone: bFM-2, MilliporeSigma™

Catalog No. 05118MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-118-MI 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-118-MI Supplier MilliporeSigma Supplier No. 05118
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect FGF-2/basic FGF using this Anti-FGF-2/basic FGF Antibody, clone bFM-2 validated for use in immunohistochemistry, RIA and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF-2/basic FGF
Applications Immunohistochemistry, Radioimmune Assays (RIA), Western Blot
Classification Monoclonal
Clone bFM-2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing PBS, pH 7.4, purified by ammonium sulfate precipitation and DEAE Sephacryl chromatography. Frozen solution.
Gene Accession No. NM_002006.3
Gene Symbols FGF2, Prostatropin, BFGF, prostatropin, HBGF-2, FGFB
Host Species Mouse
Immunogen Purified bovine brain basic FGF (FGF-2, HBGF-2). Clone bFM-2.
Purification Method Ammonium sulfate precipitation and DEAE-cellulose chromatography
Quantity 500 μg
Primary or Secondary Primary
Target Species Bovine, Human, Mouse, Rat
Content And Storage −20°C for up to 12 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.