Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human FGFR2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
$200.50 - $2171.79
Specifications
| Antigen | FGFR2 |
|---|---|
| Clone | 98742 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB6842SP
![]() |
R&D Systems
MAB6842SP |
25 μg |
Each for $200.50
|
|
|||||
MAB6842100
![]() |
R&D Systems
MAB6842100 |
100 μg |
Each for $591.67
|
|
|||||
MAB6842500
![]() |
R&D Systems
MAB6842500 |
500 μg |
Each for $2,171.79
|
|
|||||
Description
FGFR2 Monoclonal specifically detects FGFR2 in Human samples. It is validated for Western Blot.Specifications
| FGFR2 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| BBDS, BEK, BFR-1, CD332, CD332 antigen, CEK3, CFD1, craniofacial dysostosis 1, EC 2.7.10, EC 2.7.10.1, ECT1, FGF R2, fibroblast growth factor receptor 2, FLJ98662, Jackson-Weiss syndrome, JWS, Keratinocyte growth factor receptorreceptor like 14, KGFR, K-sam, KSAM, soluble FGFR4 variant 4, TK14, TK25 | |
| FGFR2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 98742 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 2263 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human FGF R2 isoforms and Mouse myeloma cell line NS0-derived recombinant human FGF R2 isoforms | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects all isoforms of human FGF R2 in direct ELISAs and Western blots. Does not cross-react with any isoform of mouse FGF R2 or human FGF R1, FGF R3, or FGF R4. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title