Learn More
FGF19, Mouse, Clone: 4C4, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant FGF19.
Supplier: Abnova Corporation H00009965M01
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes including embryonic development cell growth, morphogenesis, tissue repair, tumor growth and invasion. This growth factor is a high affinity, heparin dependent ligand for FGFR4. Expression of this gene was detected only in fetal but not adult brain tissue. Synergistic interaction of the chick homolog and Wnt-8c has been shown to be required for initiation of inner ear development. [provided by RefSeq
Sequence: MRSGCVVVHVWILAGLWLAVAGRPLAFSDAGPHVHYGWGDPIRLRHLYTSGPHGLSSCFLRIRADGVVDCARGQSAHSLLEIKAVALRTVAIKGVHSVRYLCMGADGKMQGLLQYSEEDCAFEEEIRPDGYNVYRSEKHRLPVSLSSAKQRQLYKNRGFLPLSHFLPMLPMVPEEPEDLRGHLESDMFSSPLETDSMDPFGLVTGLEAVRSPSFEKSpecifications
FGF19, | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC017664 | |
Mouse | |
Affinity chromatography | |
RUO | |
9965 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
ELISA, Western Blot | |
4C4 | |
Mouse monoclonal antibody raised against a full-length recombinant FGF19. | |
FGF19 | |
FGF19 | |
FGF19 (AAH17664, 1 a.a. ∼ 216 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.