Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-FGF8 Clone: 2A10,

Catalog No. ST1630100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ST1630100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ST1630100UG Supplier MilliporeSigma™ Supplier No. ST1630100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects FGF8 Clone: 2A10 in Human, Mouse, Rat samples, and it is validated for ELISA, Immuno Blotting

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein is known to be a factor that supports androgen and anchorage independent growth of mammary tumor cells. Overexpression of this gene has been shown to increase tumor growth and angiogensis. The adult expression of this gene is restricted to testes and ovaries. Temporal and spatial pattern of this gene expression suggests its function as an embryonic epithelial factor. Studies of the mouse and chick homologs revealed roles in midbrain and limb development, organogenesis, embryo gastrulation and left-right axis determination. The alternative splicing of this gene results in four transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGF8
Applications ELISA, Immunoblot, Immunoblot
Classification Monoclonal
Clone 2A10
Conjugate Unconjugated
Formulation In PBS, pH 7.2.
Host Species Mouse
Immunogen Full-length, recombinant, human FGF8 (aa 65-133), expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽30-35kDa FGF8 protein in PC-12, Jurkat, HepG2, NIH3T3, and Raw 264.7 cells.
Target Species Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.