Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

fibroblast growth factor binding protein 1, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016990 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-990 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-990 Supplier Abnova Corporation Supplier No. H00009982B02P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human FGFBP1 protein.

FGFBP1, or HBP17, binds to both acidic (FGF1; MIM 131220) and basic (FGF2; MIM 134920) fibroblast growth factors in a reversible manner. It also binds to perlecan (HSPG2; MIM 142461).[supplied by OMIM

Sequence: MKICSLTLLSFLLLAAQVLLVEGKKKVKNGLHSKVVSEQKDTLGNTQIKQKSRPGNKGKFVTKDQANCRWAATEQEEGISLKVECTQLDHEFSCVFAGNPTSCLKLKDERVYWKQVARNLRSQKDICRYSKTAVKTRVCRKDFPESSLKLVSSTLFGNTKPRKEKTEMSPREHIKGKETTPSSLAVTQTMATKAPECVEDPDMANQRKTALEFCGETWSSLCTFFLSIVQDTSC

Specifications

Antigen fibroblast growth factor binding protein 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human FGFBP1 protein.
Formulation PBS with no preservative; pH 7.4
Gene FGFBP1
Gene Accession No. NM_005130
Gene Alias FGFBP/HBP17
Gene Symbols FGFBP1
Host Species Mouse
Immunogen FGFBP1 (NP_005121, 1 a.a. ∼ 234 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Cycle
Primary or Secondary Primary
Gene ID (Entrez) 9982
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.