Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGFR1 Mouse anti-Human, Rat, Clone: 19B2, MilliporeSigma™

Catalog No. 05149MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-149-MI 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-149-MI Supplier MilliporeSigma Supplier No. 05149
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-FGFR1 Antibody, clone 19B2 detects level of FGFR1 and has been published and validated for use in immunoprecipitation and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FGFR1
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 19B2
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl containing 0.05% sodium azide
Gene Accession No. NM_015850.3; NM_023105.2; NM_023106.2; NM_023107.2; NM_023108.2; NM_023110.2; NM_023111.2
Gene Symbols FGFR1, CEK, bFGF-R, N-SAM, H5, FLT2, BFGFR, FGFR-1, c-fgr, FLG, H3, H2, KAL2, HBGFR, CD331, H4, FGFBR
Host Species Mouse
Immunogen E. coli fusion protein corresponding to amino acids 23-325 of human FGF receptor beta (Flg)
Purification Method Protein G chromatography
Quantity 250 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2 years at −20°C or 2 months at 4°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.