Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FGL1, Mouse, Clone: 2A4, Abnova™

Catalog No. 89022098 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-098 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-098 Supplier Abnova Corporation Supplier No. H00002267M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant FGL1.

Fibrinogen-like 1 is a member of the fibrinogen family. This protein is homologous to the carboxy terminus of the fibrinogen beta- and gamma- subunits which contains the four conserved cysteines of fibrinogens and fibrinogen related proteins. However, this protein lacks the platelet-binding site, cross-linking region and a thrombin-sensitive site which are necessary for fibrin clot formation. This protein may play a role in the development of hepatocellular carcinomas. Four alternatively spliced transcript variants encoding the same protein exist for this gene. [provided by RefSeq

Sequence: EISALEDCAQEQMRLRAQVRLLETRVKQQQVKIKQLLQENEVQFLDKGDENTVIDLGSKRQYADCSEIFNDGYKLSGFYKIKPLQSPAEFSVYCDMSDGGGWTVIQRRSDGSENFNRGWKDYENGFGNFVQKHGEYWLGNKNLHFLTTQEDYTLKIDLADFEKNSRYAQYKNFKVGDEKNFYELNIGEYSGTAGDSLAGNFHPEVQWWASHQRMKFSTWDRDHDNYEGNCAEEDQSGWWFNRCHSANLNGVYYSGPYTAKTDNGIVWYTWHGWWYSLKSVVMKIRPNDFIPNVI

Specifications

Antigen FGL1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2A4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant FGL1.
Formulation PBS with no preservative; pH 7.4
Gene FGL1
Gene Accession No. BC007047
Gene Alias HFREP1/HP-041/LFIRE1/MGC12455
Gene Symbols FGL1
Host Species Mouse
Immunogen FGL1 (AAH07047.1, 19 a.a. ∼ 312 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2267
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.