Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FHIT, Mouse, Clone: 1B5, Abnova™

Catalog No. 89022111 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-111 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-111 Supplier Abnova Corporation Supplier No. H00002272M06
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant FHIT.

This gene, a member of the histidine triad gene family, encodes a diadenosine 5',5‴-P1,P3-triphosphate hydrolase involved in purine metabolism. The gene encompasses the common fragile site FRA3B on chromosome 3, where carcinogen-induced damage can lead to translocations and aberrant transcripts of this gene. In fact, aberrant transcripts from this gene have been found in about half of all esophageal, stomach, and colon carcinomas. [provided by RefSeq]

Sequence: VVPGHVLVCPLRPVERFHDLRPDEVADLFQTTQRVGTVVEKHFHGTSLTFSMQDGPEAGQTVKHVHVHVLPRKAGDFHRNDSIYEELQKHDKEDFPASWR

Specifications

Antigen FHIT
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant FHIT.
Formulation PBS with no preservative; pH 7.4
Gene FHIT
Gene Accession No. BC032336
Gene Alias AP3Aase/FRA3B
Gene Symbols FHIT
Host Species Mouse
Immunogen FHIT (AAH32336, 31 a.a. ∼ 130 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2272
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.