Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fhl1 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABC474
Description
Detect Four and a half LIM domains protein 1 using this rabbit polyclonal antibody, Anti-Fhl1 Antibody validated for use in western blotting, immunocytochemistry and immunohistochemistry.
Specifications
Fhl1 | |
Polyclonal | |
Rabbit polyclonal in buffer containing serum with 0.05% sodium azide. | |
Rabbit | |
Unpurified | |
Primary | |
Human, Mouse, Rat | |
Serum |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
Unconjugated | |
FHL1; SLIM1 | |
Linear peptide corresponding to amino acid sequence CRDPLQGKKYVQKDGRH of human Fhl1. | |
100 μL | |
NP_001153171 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction