Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FHL2, Mouse, Clone: 2G3-1A5, Abnova™

Catalog No. 89022116 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-022-116 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-022-116 Supplier Abnova Corporation Supplier No. H00002274M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant FHL2.

LIM proteins contain a highly conserved double zinc finger motif called the LIM domain.[supplied by OMIM

Sequence: MTERFDCHHCNESLFGKKYILREESPYCVVCFETLFANTCEECGKPIGCDCKDLSYKDRHWHEACFHCSQCRNSLVDKPFAAKEDQLLCTDCYSNEYSSKCQECKKTIMPGTRKMEYKGSSWHETCFICHRCQQPIGTKSFIPKDNQNFCVPCYEKQHAMQCVQCKKPITTGGVTYREQPWHKECFVCTACRKQLSGQRFTARDDFAYCLNCFCDLYAKKCAGCTNPISGLGGTKYISFEERQWHNDCFNCKKCSLSLVGRGFLTERDDILCPDCGKDI

Specifications

Antigen FHL2
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2G3-1A5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant FHL2.
Formulation PBS with no preservative; pH 7.4
Gene FHL2
Gene Accession No. BC014397
Gene Alias AAG11/DRAL/SLIM3
Gene Symbols FHL2
Host Species Mouse
Immunogen FHL2 (AAH14397, 1 a.a. ∼ 279 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2274
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.