Learn More
FHL5, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human FHL5 peptide using ARM Technology.
Supplier: Abnova Corporation H00009457K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The protein encoded by this gene is coordinately expressed with activator of cAMP-responsive element modulator (CREM). It is associated with CREM and confers a powerful transcriptional activation function. CREM acts as a transcription factor essential for the differentiation of spermatids into mature spermatozoa. There are multiple polyadenylation sites found in this gene. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
FHL5 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
FHL5 | |
FHL5 | |
A synthetic peptide of human FHL5 | |
RUO | |
9457 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
four and a half LIM domains 5 | |
FHL5 | |
ACT, FLJ33049, KIAA0776, RP3-393D12.2, dJ393D12.2 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.