Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrillin-1 Mouse anti-Human, Clone: 69, MilliporeSigma™

Catalog No. MAB2499MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2499MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2499MI Supplier MilliporeSigma Supplier No. MAB2499
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Fibrillin-1 using this Anti-Fibrillin-1 Antibody, C-terminus, clone 69 validated for use in ELISA, immunoprecipitation, western blotting and immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrillin-1
Applications ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 69
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid at 1mg/mL in 20mM phosphate buffer, 250mM NaCl, pH 7.6, containing 0.1% sodium azide.Note: Sodium azide is toxic. MSDS available upon request.
Gene Accession No. NM_000138.3
Gene Symbols FBN1, FBN, MASS, WMS, MFS1, OCTD, SGS
Host Species Mouse
Immunogen Human Fibrillin-1
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.