Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrillin Mouse anti-Bovine, Human, Pig, Clone: 11C1.3, MilliporeSigma™

Catalog No. MAB1919MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1919MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1919MI Supplier MilliporeSigma Supplier No. MAB1919
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Fibrillin Antibody, clone 11C1.3 is an antibody against Fibrillin for use in ELISA, immunofluoresence, immunoprecipitation, western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrillin
Applications ELISA, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 11C1.3
Conjugate Unconjugated
Formulation Liquid
Gene Accession No. NM_000138.3
Gene Symbols FBN1, FBN, WMS, MFS1, OCTD, MASS, SGS
Host Species Mouse
Immunogen Bovine zonular fibrils
Quantity 100 μL
Primary or Secondary Primary
Target Species Bovine, Human, Pig
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.