Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibrinogen Sheep anti-Human, FITC, Polyclonal, MilliporeSigma™

Catalog No. AB7144FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB7144FMI 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB7144FMI Supplier MilliporeSigma Supplier No. AB7144F
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

Detect Fibrinogend using this Anti-Fibrinogen Antibody, FITC conjugated validated for use in immunofluoresence, ID and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibrinogen
Applications Immunofluorescence, Western Blot
Classification Polyclonal
Conjugate FITC
Formulation The antibody is supplied in 20mM Tris, 70mM NaCl, pH 8.0, with 0.1% sodium azide as preservative.
Gene Symbols FGA; MGC119423; MGC119425; Fib2; MGC119422
Host Species Sheep
Immunogen Purified fibrinogen derived from pooled human plasma.
Quantity 2 mL
Primary or Secondary Primary
Gene ID (Entrez) NM_000508.3; NM_021871.2
Target Species Human
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months. Protect from light.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.