Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Goat IgG Fraction to Human Fibrinogen, FITC, Polyclonal, MP Biomedicals
Supplier: MP Biomedicals, Inc 0855169
Description
Host Species: Goat
Species Reactivity: Human

Specifications
Fibrinogen | |
FITC | |
2 mL | |
Human |
Polyclonal | |
Goat | |
Primary | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction