Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibroblast activation protein alpha (FAP) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABT11MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT11MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT11MI Supplier MilliporeSigma Supplier No. ABT11
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Fibroblast activation protein alpha (FAP) using this Anti-Fibroblast activation protein alpha (FAP) Antibody validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibroblast activation protein alpha (FAP)
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing PBS (pH 7.2) with 0.05% sodium azide.
Gene Symbols FAPA; FAP; DPPIV
Host Species Rabbit
Immunogen Histidine-tagged recombinant protein corresponding to human FAP.
Purification Method Ammonium Sulfate
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_004451
Target Species Human
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.