Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibroblast Growth Factor Receptor 1 Mouse anti-Human, Mouse, Rat, Chicken, Bovine, Clone: VBS1, MilliporeSigma™

Catalog No. 051134MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-113-4MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-113-4MI Supplier MilliporeSigma Supplier No. 051134
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Fibroblast Growth Factor Receptor 1 using this Anti-Fibroblast Growth Factor Receptor 1 Antibody, clone VBS1 validated for use in western blot, immunoprecipitation, immunohistochemistry (paraffin), ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibroblast Growth Factor Receptor 1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone VBS1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgM in 0.015Mpotassium phosphate buffer containing 0.15MNaCl, 0.1% sodium azide, pH 7.20.
Gene Accession No. NP_001103677.1
Gene Symbols CD331, CEK, EC 2.7.10.1, FGFBR, FGFR-1, FLG, FLJ99988, FLT2, H2, H3, H4, H5, HBGFR, KAL2, N-SAM , OGD, OTTHUMP00000190874, OTTHUMP00000190878 , OTTHUMP00000190879, OTTHUMP00000190881, bFGF-R, c-fgr , BFGFR
Host Species Mouse
Immunogen FGF Receptor purified from bovine coronary venular endothelial cells.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Chicken, Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.