Learn More
Fibronectin Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB2040
Description
Anti-Fibronectin Antibody is an antibody against Fibronectin for use in ELISA, RIA, immunohistochemistry.
Specifications
Fibronectin | |
Polyclonal | |
IgG fraction. No preservatives. Liquid containing 0.01M Phosphate, 0.09M NaCl, pH7.2. | |
Rabbit | |
100 μg | |
NM_002026.2; NM_054034.2; NM_212474.1; NM_212475.1; NM_212476.1; NM_212478.1; NM_212482.1 | |
−20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles. |
ELISA, Immunohistochemistry, Radioimmune Assays (RIA) | |
Unconjugated | |
FN1; DKFZp686O13149; DKFZp686H0342; DKFZp686I1370; MSF; FN; CIG; DKFZp686F10164; LETS; FINC | |
Fibronectin extracted and purified from rat plasma. | |
Primary | |
Rat | |