Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB2033MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2033MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2033MI Supplier MilliporeSigma Supplier No. AB2033
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Fibronectin using this Anti-Fibronectin Antibody validated for use in ELISA, immunocytochemistry, immunohistochemistry, RIA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibronectin
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Radioimmune Assays (RIA)
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.01M phosphate, 0.09M NaCl, pH 7.2 with no preservatives.
Gene Symbols FN1; MSF; FN; CIG; LETS; FINC
Host Species Rabbit
Immunogen Fibronectin extracted and purified from mouse plasma.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_002026.2
Target Species Mouse
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.