Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fibronectin (Cell Attachment Fragment) Mouse anti-Human, Clone: 3E3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB88916C
Description
This Anti-Fibronectin (Cell Attachment Fragment) Antibody, clone 3E3, Ascites and Azide Free is validated for use in western blotting, immunohistochemistry, ELISA, immunocytochemistry and inhibition for the detection of Fibronectin.
Specifications
Fibronectin (Cell Attachment Fragment) | |
Monoclonal | |
Unconjugated | |
NP_002017 | |
Mouse | |
Protein G purified | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 κ |
ELISA, Immunocytochemistry, Immunohistochemistry, Inhibition Assays, Western Blot | |
3E3 | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl without preservatives. | |
FN1, FN | |
Purified human Fibronectin, which contains the Cell Attachment Fragment, from amnion liquid. | |
100 μg | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction