Learn More
MilliporeSigma™ anti-Fibronectin, cell binding domain, Clone: 568, ™ (Chemicon™)
Supplier: MilliporeSigma™ MAB1937/DEL
Description
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Radioimmunoassay, Western Blotting
Specifications
Fibronectin, cell binding domain | |
Unconjugated | |
Proteins secreted by cultured human embryonic fibroblasts. | |
RUO | |
Primary | |
Ascites |
Monoclonal | |
Mouse | |
100 μL | |
Cell Structure | |
Human | |
IgG1 |