Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Mouse, Unlabeled, Clone: 10, BD

Catalog No. BDB610078 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610078 150 μg
BDB610077 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610078 Supplier BD Biosciences Supplier No. 610078
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 240kDa dimeric fibronectin protein exists in two forms: a soluble protomer in body fluids and an insoluble multimer in the extracellular matrix. The latter is the primary functional form and creates a substrate for cell migration, a role which makes fibronectin vital to embryogenesis and wound response. Fibronectin mediates cytoskeletal organization, cell attachment, and cellular signaling through interactions with cellular receptors. Although various isoforms of fibronectin are derived by alternative splicing, they share a common N-terminus which is a critical region for cell surface binding in an initial step of multimer assembly. Further polymerization steps are regulated by fibronectin/integrin interactions and result in generation of the complex fibrils that constitute the fibronectin matrix.

Host Species: Mouse
Clone: 10
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Fibronectin
Formula Weight [Chemical]: 240kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibronectin
Applications Flow Cytometry
Classification Monoclonal
Clone 10
Concentration 250μg/mL
Conjugate Unconjugated
Description FN; LETS
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Fibronectin
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Bovine, Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.