Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Mouse anti-Human, Rat, FITC, Clone: 5G7, Southern Biotech™
SDP

Catalog No. OB147002 Shop All Southern Biotechnology Associates Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OB147002 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OB147002 Supplier Southern Biotechnology Associates Supplier No. 147002
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SouthernBiotech is a leading provider of antibodies and proteins for extracellular matrix research.

Specifications

Antigen Fibronectin
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 5G7
Concentration 0.1mg/mL
Conjugate FITC
Formulation Supplied in PBS/NaN3
Host Species Mouse
Immunogen Human plasma fibronectin
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store at 2-8°C; Protect from light
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.