Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fibronectin Mouse anti-Human, Clone: IST-10, MilliporeSigma™

Catalog No. MAB1892MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1892MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1892MI Supplier MilliporeSigma Supplier No. MAB1892
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Fibronectin Antibody, gelatin binding domain, clone IST-10 is an antibody against Fibronectin for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fibronectin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone IST-10
Conjugate Unconjugated
Formulation Purified immunoglobulin in 0.02M phosphate buffer, 0.25M sodium chloride, 0.1% sodium azide, pH 7.6
Gene Accession No. NM_002026.2; NM_054034.2; NM_212474.1; NM_212475.1; NM_212476.1; NM_212478.1; NM_212482.1
Gene Symbols FN1, DKFZp686H0342, DKFZp686I1370, MSF, CIG, LETS, DKFZp686O13149, FN, DKFZp686F10164, FINC
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.