Learn More
FIGLA, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human FIGLA peptide using ARM Technology.
Supplier: Abnova Corporation H00344018K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a protein that functions in postnatal oocyte-specific gene expression. The protein is a basic helix-loop-helix transcription factor that regulates multiple oocyte-specific genes, including genes involved in folliculogenesis and those that encode the zona pellucida. Mutations in this gene cause premature ovarian failure type 6. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
FIGLA | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
FIGLA | |
FIGLA | |
A synthetic peptide of human FIGLA | |
RUO | |
344018 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
folliculogenesis specific basic helix-loop-helix | |
FIGLA | |
POF6, bHLHc8 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.