Learn More
Description
Specifications
Specifications
| Antigen | Filamin B |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in 0.02M PBS, 0.25M NaCl containing 0.1% sodium azide. |
| Gene Symbols | FLNB; FH1; FLN3; LRS1; AOI; DKFZp686A1668; ABP-278; Filamin-B; SCT; Fh1; FLN1L; TAP; TABP; FLN-B; Beta-filamin; DKFZp686O033 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide from human filamin B. The immunogen peptide corresponds to a sequence near the hinge 2 region of Filamin B (near C-term) and is not present in the two C-terminally truncated splice variants of Filamin B (Var-2 and Var-3). The peptide sho |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.