Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FIP200 Rabbit anti-Human, Rat, Mouse, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma 071815
Description
Detect FIP200 using this Anti-FIP200 Antibody validated for use in western blotting.
Specifications
FIP200 | |
Polyclonal | |
Unpurified rabbit serum in buffer containing 0.05% sodium azide. | |
The immunogen was a synthetic peptide corresponding to the C-terminal region of FIP200. | |
Primary | |
Human, Mouse, Rat | |
Serum |
Western Blot | |
Unconjugated | |
Rabbit | |
100 μL | |
Q86YR4 | |
-80°C in undiluted aliquots for up to 24 months, Avoid Freeze/Thaw Cycles |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction