Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP-12 Mouse, Unlabeled, Clone: 2C1-87, BD

Catalog No. BDB554091 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554091 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554091 Supplier BD Biosciences Supplier No. 554091

Mouse Monoclonal Antibody

FKBP-12 (for FK506 binding protein-12kDa) is a ubiquitously expressed protein involved in T-cell signaling that binds to FK506. FK506 belongs to a class of proteins that have peptidyl-prolyl cis-trans isomerase (PPIase) activity. This class of proteins is thought to be involved in folding proteins and peptides into their native conformations. It is thought that peptidyl-prolyl isomerization may play a role in regulating components of the T-cell signal transduction pathway such as transcription factors, protein kinase activation, and ion channel function.
FK506, like cyclosporin, is an agent that inhibits T- and B-cell early activation processes that are key to an effective immune response. In T cells these agents disrupt a step in the signal transduction pathway from the T-cell receptor to cytokine genes involved in coordinating the immune response. In particular, they seem to disrupt pathways that induce a measurable rise in Ca2+. NFAT, a transcription factor essential for early T-cell activation, appears to be a specific target of FK506 and cyclosporin because transcription directed by NF-AT is specifically blocked in T cells treated with these agents. NFAT is formed when a signal from the T-cell antigen receptor induces a pre-existing cytoplasmic subunit to translocate to the nucleus and combine with a newly synthesized nuclear subunit of NFAT. When FK506 binds to FKBP-12, the complex blocks translocation of the cytoplasmic subunit of NF-AT into the nucleus by inhibiting a Ca2+-dependent phosphatase. Since the cytoplasmic subunit is not translocated into the nuclear, the formation of NFAT and its subsequent transcriptional activity is blocked. The reduced molecular weight of FKBP-12 is 12kDa. The 2C1-87 antibody recognizes FKBP-12. Specifically it has been shown to recognize human, mouse, and rat FKBP-12. Recombinant FKBP-12 was used as immunogen. The antibody was originally characterized by biochemical, immunohistochemical, and functional assays.

Host Species: Mouse
Clone: 2C1-87
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant FKBP-12
Formula Weight [Chemical]: 12kDa

Immunohistochemistry (Paraffin), Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen FKBP-12
Applications Western Blot
Classification Monoclonal
Clone 2C1-87
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Recombinant FKBP-12
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.