Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FKBP5 Matched Antibody Pair, Abnova

Catalog No. 89017279 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-279 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-279 Supplier Abnova Corporation Supplier No. H00002289AP11
Add to Cart
Add to Cart

Includes

  • 1. Capture antibody: rabbit affinity purified polyclonal anti-FKB5 (100μg)
  • 2. Detection antibody: mouse monoclonal anti-FKBP5 κ (20μg)
  • Sufficient for at least 3-5 x 96 well plates using recommended protocols.

The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This encoded protein is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin. It is thought to mediate calcineurin inhibition. It also interacts functionally with mature hetero-oligomeric progesterone receptor complexes along with the 90 kDa heat shock protein and P23 protein. This gene has been found to have multiple polyadenylation sites.

  • Species Reactivity: Human

ELISA

Specifications

Antigen FKBP5
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FKBP5.
Gene FKBP5
Gene Alias FKBP51/FKBP54/MGC111006/P54/PPIase/Ptg-10
Gene Symbols FKBP5
Host Species Mouse, Rabbit
Immunogen NA; FKBP5 (AAH42605, 1 a.a. ∽ 458 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2289
Includes Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FKBP5 (100 μg) 2. Detection antibody: mouse monoclonal anti-FKBP5, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.