Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLI-1 Mouse anti-Human, Unlabeled, Clone: G146-254, BD

Catalog No. BDB554267 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB554267 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB554267 Supplier BD Biosciences Supplier No. 554267
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Fli-1 is a 50kDa ets-related transcription factor. Like other ets-related proteins it binds to consensus sites in DNA through a 85 amino acid ets domain in the carboxyl region of the protein. Fli-1 is associated with Ewing sarcoma through at (11;22)(q24;q12) chromosomal translocation. This translocation brings the 5' region of the EWS gene into conjunction with the 3' region of the Fli-1 gene encoding the ets-DNA binding domain. Such a translocation is found in 85% of Ewing sarcomas. The resulting fusion protein has the DNA binding activity of Fli-1 and, with the EWS domain, it is also a more potent transcriptional activator than Fli-1 itself. The strong transforming activity of the EWS-Fli-1 fusion protein may in part be due to its ability to trans-activate the promoter for c-myc. C-myc is known to be elevated in Ewing sarcomas. Clone G146-254 was raised against a bacterially expressed Fli-1 ets domain fusion protein.

Host Species: Mouse
Clone: G146-254
Isotype: IgG2a
Species Reactivity: Human
Immunogen: Fli-1 ets Domain Fusion Protein
Formula Weight [Chemical]: 50kDa

Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen FLI-1
Applications Western Blot
Classification Monoclonal
Clone G146-254
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Fli-1 ets Domain Fusion Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.