Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-FLJ13910, Polyclonal, Abnova

Catalog No. 89008101 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-101 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-101 Supplier Abnova Corporation Supplier No. H00064795B01

Mouse Polyclonal

Detection Antibody, Western Blotting

Specifications

Antigen required for meiotic nuclear division 5 homolog A (S. cerevisiae)
Classification Polyclonal
Conjugate Unconjugated
Host Species Mouse
Immunogen FLJ13910 (AAH12165, 1 a.a. ∽392 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Stem Cell Biology
Whole Molecule Yes
Primary or Secondary Primary
Target Species Human
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.