Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FAST kinase domains 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant FLJ21901.
Supplier: Abnova Corporation H00079675A01
Description
Sequence: KKTPVFLESLVTNMLRLRAICPFSWRVFQFRPISCEPLIIQMNKCTDEEQMFGFIERNKAILSEKQVGCAFDMLWKLQKQKTSLLKNAEYVRDHPQFLSpecifications
FAST kinase domains 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant FLJ21901. | |
FASTKD1 | |
FLJ21901/KIAA1800 | |
Mouse | |
50 μL | |
Stem Cell Biology | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_024622 | |
FASTKD1 | |
FLJ21901 (NP_078898, 2 a.a. ∼ 99 a.a) partial recombinant protein with GST tag. | |
RUO | |
Yes | |
79675 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction