Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

leucine-rich repeat, immunoglobulin-like and transmembrane domains 3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008744 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-744 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-744 Supplier Abnova Corporation Supplier No. H00345193B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human LRIT3 protein.

Sequence: MPLLRTLDLHNNKITSVPNEALRYLKNLAYLDLSSNRLTTLTPDFLENWTHLVSTPSGVLDLSPSRIILGLQDNPWFCDCHISKMIELSKVVDPAIVLLDPLMTCSEPERLTGILFQRAELEHCLKPSVMTSATKIMSALGSNVLLRCDATGFPTPQITWTRSDSSPVNYTVIQESPEEGVRWSIMSLTGISSKDAGDYKCKAKNLAGMSEAVVTVTVLGITTTPIPPDTSERTGDHPEWDVQPGSGRSTSVSSASSYLWSSSFSPTSSFSASTLSPPSTASFSLSPFSSSTVSSTTTLSTSISASTTMANKRSFQLHQGGKRNLKVAKNGSKLPPASTSKKEELALLDQTMLTETNATIENLRVVSETKESVTLMWNMINTTHNSAVTVLYSKYGGKDLLLLNADSSKNQVTIDGLEPGGQYMACVCPKGVPPQKDQCITFSTERVEGDDSQWSLLLVVTSTACVVILPLICFLLYKVCKLQCKSEPFWEDDLAKETYIQFETLFPRSQSVGELWTRSHRDDSEKLLLCSRSSVESQVTFKSEGSRPEYYC

Specifications

Antigen leucine-rich repeat, immunoglobulin-like and transmembrane domains 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human LRIT3 protein.
Formulation No additive
Gene LRIT3
Gene Accession No. BC104037.1
Gene Alias FIGLER4/FLJ44691/MGC120618
Gene Symbols LRIT3
Host Species Mouse
Immunogen LRIT3 (AAI04038.1, 1 a.a. ∼ 552 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Adhesion
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 345193
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.