Learn More
Flk-1 Rat anti-Mouse, PE-Cy7, Clone: Avas 12alpha1, BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 561259
Description
The Avas 12α1 monoclonal antibody specifically binds to fetal liver kinase 1 (Flk-1), a receptor protein tyrosine kinase closely related to CD117 (c-kit) and CD140a (PDGF Receptor α chain) of the immunoglobulin superfamily. Flk-1, also known as VEGF Receptor-2 (VEGF-R2), is a receptor for vascular endothelial growth factor (VEGF). It is expressed, at the mRNA and protein levels, on distinct sets of mesoderm during gastrulation and on endothelial cells in embryonic and adult tissues. In vivo and in vitro studies indicate that Flk-1 is required for the embryonic development of vascular endothelial and hematopoietic cells.

Specifications
Flk-1 | |
Monoclonal | |
0.2mg/mL | |
Fetal liver kinase 1; CD309; Kdr; VEGF receptor-2; VEGFR-2 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
Avas 12alpha1 | |
PE-Cyanine7 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Flk-1 Recombinant Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.