Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Flt-3/Flk-2, Alexa Fluor 700, Clone: 66903, R&D Systems™

Catalog No. FAB812N
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB812N 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB812N Supplier R&D Systems Supplier No. FAB812N
This item is not returnable. View return policy

Applications:

Flow Cytometry

Specifications

Antigen Flt-3/Flk-2/CD135
Applications Flow Cytometry
Classification Monoclonal
Clone 66903
Conjugate Alexa Fluor 700
Dilution Flow Cytometry 5 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Accession No. AAA18947
Gene Alias CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, Flk-2, FLK2, Flt3, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Gene Symbols FLT3
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human Flt-3/Flk-2, Asn27-Asn541, Accession # AAA18947
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2322
Test Specificity Detect human Flt-3/Flk-2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Flt-3, recombinant human (rh) SCF R, rhPDGF R alpha , or rhPDGF R beta is observed.
Target Species Human
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 °C as supplied.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.