Learn More
FLT4, Mouse, Clone: 4D1, Abnova™
Mouse monoclonal antibody raised against a partial recombinant FLT4.
Supplier: Abnova Corporation H00002324M05
Description
This gene encodes a tyrosine kinase receptor for vascular endothelial growth factors C and D. The protein is thought to be involved in lymphangiogenesis and maintenance of the lymphatic endothelium. Mutations in this gene cause hereditary lymphedema type IA. [provided by RefSeq
Sequence: ITEESHVIDTGDSLSISCRGQHPLEWAWPGAQEAPATGDKDSEDTGVVRDCEGTDARPYCKVLLLHEVHANDTGSYVCYYKYIKARIEGTTAASSYVFVRSpecifications
FLT4 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_002020 | |
FLT4 | |
FLT4 (NP_002011, 34 a.a. ∼ 133 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
4D1 | |
Mouse monoclonal antibody raised against a partial recombinant FLT4. | |
FLT4 | |
FLT41/LMPH1A/PCL/VEGFR3 | |
Mouse | |
Affinity chromatography | |
RUO | |
2324 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.