Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein Mouse anti-All, AlexaFluor™488, Clone: 5D6.2, MilliporeSigma™

Catalog No. MAB045XMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB045XMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB045XMI Supplier MilliporeSigma Supplier No. MAB045X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Fluorescein using this Anti-Fluorescein Antibody, clone 5D6.2, Alexa Fluor 488 conjugated validated for use in immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fluorescein
Applications Immunofluorescence
Classification Monoclonal
Clone 5D6.2
Conjugate Alexa Fluor 488
Formulation Purified immunoglobulin conjugated to Alexa Fluor™ 488. Liquid in Phosphate buffer with 15mg/mL BSA as a stabilizer and 0.1% sodium azide.
Host Species Mouse
Immunogen Fluorescein-KLH
Quantity 100 μg
Primary or Secondary Primary
Target Species All species
Content And Storage 2-8°C in the dark for up to 6 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.