Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein (FITC) Mouse anti-All, HRP, Clone: 5D6.2, MilliporeSigma™

Catalog No. MAB045PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB045PMI 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB045PMI Supplier MilliporeSigma Supplier No. MAB045P
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Fluorescein (FITC) using this Anti-Fluorescein (FITC) Antibody, clone 5D6.2, Peroxidase conjugated validated for use in ELISA, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Fluorescein
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone 5D6.2
Conjugate HRP
Formulation Lyophilized from 0.01M PBS, containing 0.1% Thimerosal and 15mg/mL BSA. Reconstitute with 1 mL of sterile distilled water.
Host Species Mouse
Quantity 1 mL
Primary or Secondary Primary
Target Species All species
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Lyophilized
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.