Learn More
Description
Host Species: Mouse
Clone: F4/1
Isotype: IgG1/κ
Cross Reactivity: All species
Immunogen: Fluorescein isothiocyanate (FITC)
Ab-1 recognizes both the free and protein-conjugated (either soluble or cell bound) form of fluorescein or carboxyfluorescein. Sensitivity of the FITC-anti-FITC system is reportedly similar to that of the biotin-avidin system. In biotin or enzyme-labeled form, NeoMarkers' Ab-1 is also suitable for the conversion of fluorescein-labeled detection into immuno-enzymatic assays.
Flow Cytometry, Immunohistochemistry
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
| Antigen | Fluorescein Isothiocyanate (FITC) Ab-1 |
| Classification | Monoclonal |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Fluorescein isothiocyanate (FITC) |
| Quantity | 1 mL |
| Primary or Secondary | Primary |
| Target Species | All species |
| Form | Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.