Learn More
Description
FMC7 recognizes a 105kDa membrane glycoprotein expressed on a subset of B lymphocytes.More than 50% of the peripheral B lymphocytes of normal adults carry the FMC7 antigen at variable density. FMC7-positive B cells are more mature, and they include the subpopulation that responds in vitro to mitogens or antigens. The FMC7 antigen is found on B-cell malignancies of most differentiated stages, such as mantle cell lymphoma, but not in most cases of chronic lymphocytic leukemia (CLL).
Host Species: Mouse
Clone: FMC7
Isotype: IgM, κ
Cross Reactivity: Human
Flow Cytometry
Specifications
Specifications
| Antigen | FMC7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | FMC7 |
| Concentration | 100μg/mL |
| Conjugate | Violet Fluor 450 (V450) |
| Formulation | HEPES buffer with gelatin and 0.1% sodium azide. |
| Host Species | Mouse |
| Immunogen | Human B-lymphoblastoid cell line HRIK |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.