Learn More
FMOD, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human FMOD peptide using ARM Technology.
Supplier: Abnova Corporation H00002331K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
Fibromodulin is a member of a family of small interstitial proteoglycans, containing a central region composed of leucine-rich repeats with 4 keratan sulfate chains flanked by disulfide-bonded terminal domains. It may participate in the assembly of the extracellular matrix as it interacts with type I and type II collagen fibrils and inhibits fibrillogenesis in vitro. It may also regulate TGF-beta activities by sequestering TGF-beta into the extracellular matrix. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
FMOD | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
FMOD | |
FMOD | |
A synthetic peptide of human FMOD | |
RUO | |
2331 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
fibromodulin | |
FMOD | |
SLRR2E | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.