Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMR1 Mouse anti-Human, Clone: 7H3.2, MilliporeSigma™

Catalog No. 051235MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-123-5MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-123-5MI Supplier MilliporeSigma Supplier No. 051235

Mouse Monoclonal Antibody

Anti-FMR1 Antibody, clone 7H3.2 is a Mouse Monoclonal Antibody for detection of FMR1 also known as Fragile X mental retardation 1 protein, FMRP and has been validated in western blot, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FMR1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 7H3.2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_001172004
Gene Symbols FMR1
Host Species Mouse
Immunogen Linear peptide corresponding to human FMR1.
Purification Method Ion Exchange
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.