Learn More
FAU, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human FAU peptide using ARM Technology.
Supplier: Abnova Corporation H00002197K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene is the cellular homolog of the fox sequence in the Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV). It encodes a fusion protein consisting of the ubiquitin-like protein fubi at the N terminus and ribosomal protein S30 at the C terminus. It has been proposed that the fusion protein is post-translationally processed to generate free fubi and free ribosomal protein S30. Fubi is a member of the ubiquitin family, and ribosomal protein S30 belongs to the S30E family of ribosomal proteins. Whereas the function of fubi is currently unknown, ribosomal protein S30 is a component of the 40S subunit of the cytoplasmic ribosome. Pseudogenes derived from this gene are present in the genome. Similar to ribosomal protein S30, ribosomal proteins S27a and L40 are synthesized as fusion proteins with ubiquitin. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
FAU | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
FAU | |
FAU | |
A synthetic peptide of human FAU | |
RUO | |
2197 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
Finkel-Biskis-Reilly murine sarcoma virus (FBR-MuSV) ubiquitously expressed | |
FAU | |
FAU1, FLJ22986, Fub1, Fubi, MNSFbeta, RPS30 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.